Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRG4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.50 - $353.00
Specifications
Antigen | NRG4 |
---|---|
Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB186691
![]() |
Novus Biologicals
NBP31281150UL |
50 μg |
Each for $182.50
|
|
|||||
NB186689
![]() |
Novus Biologicals
NBP312811100UL |
100 μg |
Each for $274.50
|
|
|||||
NB186690
![]() |
Novus Biologicals
NBP312811200UL |
200 μg |
Each for $353.00
|
|
|||||
Description
NRG4 Polyclonal antibody specifically detects NRG4 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
NRG4 | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Neuroscience, Signal Transduction | |
0.2μm filtered solution in PBS without Preservative | |
145957 | |
IgG | |
Protein A purified |
ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4 | |
Purified, recombinant Human NRG4 (Q8WWG1; Pro2-Phe62) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title