Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIP Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP13075A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP13075A594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP13075A594 Supplier Novus Biologicals Supplier No. NBP130075AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRIP Polyclonal specifically detects NRIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NRIP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 1200006M05Rik, Androgen receptor complex-associated protein, ARCAP, DDB1 and CUL4 associated factor 6, DDB1- and CUL4-associated factor 6, FLJ23798, IQ motif and WD repeat-containing protein 1, IQ motif and WD repeats 1, IQWD1, MSTP055, NRIP, Nuclear receptor interaction protein, PC326
Gene Symbols DCAF6
Host Species Rabbit
Immunogen Maps to a region between residue 300 and 350 of human nuclear receptor interaction protein (IQ motif and WD repeats 1) using the numbering given in entry NP_001017977.1
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55827.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.