Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIP Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP233208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233208 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233208 Supplier Novus Biologicals Supplier No. NBP233208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRIP Polyclonal specifically detects NRIP in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.

Specifications

Antigen NRIP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation 0.2 ml PBS with No Preservative
Gene Accession No. Q58WW2
Gene Alias 1200006M05Rik, Androgen receptor complex-associated protein, ARCAP, DDB1 and CUL4 associated factor 6, DDB1- and CUL4-associated factor 6, FLJ23798, IQ motif and WD repeat-containing protein 1, IQ motif and WD repeats 1, IQWD1, MSTP055, NRIP, Nuclear receptor interaction protein, PC326
Gene Symbols DCAF6
Host Species Rabbit
Immunogen A portion of amino acids 800-850 of human NRIP was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55827
Test Specificity The amino acid sequence used is 100% homologous in human (isoforms a and b), mouse, rat and dog, 88% homologous in zebrafish and 82% homologous in xenopus.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.