Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIP3 Antibody (OTI1A9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273066A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273066A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273066A30 Supplier Novus Biologicals Supplier No. NBP273066AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRIP3 Monoclonal specifically detects NRIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NRIP3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A9
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C11orf14, chromosome 11 open reading frame 14, nuclear receptor interacting protein 3, nuclear receptor-interacting protein 3, NY-SAR-105, Sarcoma antigen NY-SAR-105
Gene Symbols NRIP3
Host Species Mouse
Immunogen Full length recombinant protein of human NRIP3 (NP_065696) produced in HEK293T cells.
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56675
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.