Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRK Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB335582 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335582 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335582 Supplier Novus Biologicals Supplier No. NBP298072H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRK Polyclonal antibody specifically detects NRK in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NRK
Applications Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DKFZp686A17109, EC 2.7.11, EC 2.7.11.1, FLJ16788, MGC131849, NESK, Nik related kinase, nik-related protein kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NRK fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 203447
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.