Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRSF Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30379820UL
Description
NRSF Polyclonal specifically detects NRSF in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NRSF | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
Recombinant fusion protein containing a sequence corresponding to amino acids 760-1060 of human NRSF (NP_001180437.1). | |
20 μL | |
Stem Cell Markers, Transcription Factors and Regulators | |
5978 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction