Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRSN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP231844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231844 0.1 mL
NB437172 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231844 Supplier Novus Biologicals Supplier No. NBP231844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NRSN1 Polyclonal specifically detects NRSN1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NRSN1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8IZ57
Gene Alias neurensin 1, neurensin-1, Neuro-p24, p24, vesicular membrain protein p24, Vesicular membrane protein of 24 kDa, Vesicular membrane protein p24neuro-p24, VMP
Gene Symbols NRSN1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VKSYSKEEKFLQQKFKERIADIKAHTQPVTKAPGPGETKIPVTLSRVQNVQPLLAT
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140767
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.