Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 11021
GREENER_CHOICE

Catalog No. p-4816584
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Activator of BKCa (KCa1.1) channels

Activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1). Exhibits no modulatory effect on a variety of K+ (KV), Na+ and Ca2+ currents at concentrations <10 μM. Alters gating kinetics, but does not affect single channel conductance. Shown to bind BKCa in open and closed conformations; thought to bind the α subunit.

Chemical Identifiers

CAS 956014-19-0
Molecular Formula C16H9BrF6N6S

Specifications

Quantity 10 mg
Formula Weight 511.24
Purity >98%
Molecular Weight (g/mol) 511.244
Chemical Name or Material NS 11021
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.