Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 1643
GREENER_CHOICE

Catalog No. 306210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-6210 10 mg
30-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-6210 Supplier Tocris Bioscience™ Supplier No. 3062/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV11.1 (hERG) channel activator; antiarrhythmic

Human ether-a-go-go related gene (hERG) KV11.1 channel activator (EC50 = 10.5 μM). Exhibits different molecular mechanisms of action at KV11.1 (hERG1) and KV11.2 (hERG2) channels. Displays antiarrhythmic activity in vitro.

Chemical Identifiers

CAS 448895-37-2
Molecular Formula C15H10F6N2O3

Specifications

Quantity 10 mg
Formula Weight 380.24
Purity >98%
Molecular Weight (g/mol) 380.246
Chemical Name or Material NS 1643
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.