Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 1738
GREENER_CHOICE

Catalog No. p-4784211
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChR; active in vivo

Selective positive allosteric modulator of α7 nicotinic acetylcholine receptors. Exhibits no substantial activity for α4β2, α3β3 and α1-containing receptors. Displays cognitive-enhancing properties in vivo.

Chemical Identifiers

CAS 501684-93-1
Molecular Formula C14H9Cl2F3N2O2

Specifications

Quantity 10 mg
Formula Weight 365.13
Purity 98%
Molecular Weight (g/mol) 365.133
Chemical Name or Material NS 1738
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.