Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 2028
GREENER_CHOICE

Catalog No. 451710
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent soluble guanylyl cyclase (sGC) inhibitor

Potent soluble guanylyl cyclase (sGC) inhibitor (Ki = 8 nM). Blocks sGC activity in murine cerebellum induced by S-nitroso-glutathione and NMDA (IC50 values are 17 and 20 nM respectively). Inhibits VEGF-induced cGMP accumulation; abolishes VEGF-induced migration in postcapillary venular endothelial cells (CVEC).

Chemical Identifiers

CAS 204326-43-2
Molecular Formula C9H5BrN2O3

Specifications

Quantity 10 mg
Formula Weight 269.05
Purity >98%
Molecular Weight (g/mol) 269.05
Chemical Name or Material NS 2028
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.