Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NS 3623
GSA_VAGREENER_CHOICE

Catalog No. p-4816611
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

KV11.1 (hERG) and KV4.3 channel activator; antiarrhythmic

KV11.1 (hERG) and KV4.3 channel activator. Activates the IKr and Ito currents and displays antiarrhythmic activity. May also act as a partial blocker of KV11.1 channels. Displays selectivity over the key cardiac potassium channels, KV7.1 (KCNQ1) and KV1.5. Suitable for both in vitro and in vivo use.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 343630-41-1
Molecular Formula C15H10BrF3N6O

Specifications

Quantity 10 mg
Formula Weight 427.18
Purity >99%
Molecular Weight (g/mol) 427.185
Chemical Name or Material NS 3623
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.