Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NS 398
Cyclooxygenase-2 (COX-2) inhibitor
$276.08 - $1151.88
Specifications
Chemical Name or Material | N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide |
---|---|
CAS | 123653-11-2 |
Assay Percent Range | >99% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C13H18N2O5S |
Description
Selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.Specifications
N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide | |
>99% | |
C13H18N2O5S |
123653-11-2 | |
Soluble to 100mM in DMSO | |
Observed MW: 314.36 |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title