Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NSC 23766

Click to view available options
Quantity:
10 mg
50 mg
Description
NSC 23766 is a selective inhibitor of Rac1-GEF interaction. NSC 23766 prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∼ 50 μM) without affecting Cdc42 or RhoA activation. The compound inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells. NSC 23766 also inhibits mammary carcinoma cell invasion in vitro and inhibits mammary tumor progression and metastasis in a mouse model.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | NSC 23766 |
| Purity | >98% |
| Molecular Weight (g/mol) | 530.97 |
| Inhibitors | Rho GTPase Inhibitors |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction