Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSD3 Antibody (7B4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP104324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104324 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104324 Supplier Novus Biologicals Supplier No. NBP104324
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NSD3 Monoclonal specifically detects NSD3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen NSD3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7B4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:3000, ELISA 1:100-1:2000
Gene Accession No. NP_075447
Gene Alias EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1
Gene Symbols WHSC1L1
Host Species Mouse
Immunogen Recombinant human NSD3 (383-660aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54904
Test Specificity Anti-human NSD3 mAb, clone 2E9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NSD3 protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.