Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSDHL Antibody (20F2), Novus Biologicals™
SDP

Catalog No. NB298062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298062 50 μL
NB298063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298062 Supplier Novus Biologicals Supplier No. NBP32645750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NSDHL Monoclonal antibody specifically detects NSDHL in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen NSDHL
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 20F2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 1.1.1.170, H105e3, member 1, NAD(P) dependent steroid dehydrogenase-like, SDR31E1, sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating, XAP104
Host Species Rabbit
Immunogen A synthesized peptide derived from Human NSDHL [UniProt Q15738]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 50814
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.