Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSDHL Antibody (20F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | NSDHL |
---|---|
Clone | 20F2 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
NSDHL Monoclonal antibody specifically detects NSDHL in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
NSDHL | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.1.1.170, H105e3, member 1, NAD(P) dependent steroid dehydrogenase-like, SDR31E1, sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating, XAP104 | |
A synthesized peptide derived from Human NSDHL [UniProt Q15738] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
20F2 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
metabolism | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
50814 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title