Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Antibody (9G7-3), Novus Biologicals™
SDP

Catalog No. NBP119333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119333 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119333 Supplier Novus Biologicals Supplier No. NBP119333
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NSF Monoclonal specifically detects NSF in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NSF
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 9G7-3
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation In PBS, pH 7.2, containing 50% glycerol. with 0.09% Sodium Azide
Gene Alias EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF
Gene Symbols NSF
Host Species Mouse
Immunogen Recombinant bovine NSF protein
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4905
Test Specificity NSF (9G7-3)
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.