Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSF Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018579 0.025 mg
NBP145746 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018579 Supplier Novus Biologicals Supplier No. NBP1457460.025MG

Rabbit Polyclonal Antibody

NSF Polyclonal specifically detects NSF in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen NSF
Applications Immunoprecipitation, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Immunohistochemistry 10 - 20 ug/mL, Immunoprecipitation 5 - 15 ug/mL
Formulation PBS, 1% BSA, 50% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9QUL6
Gene Alias EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF
Gene Symbols NSF
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids around 731 of rat NSF.
Molecular Weight of Antigen 83 kDa
Purification Method Affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4905
Test Specificity The antibody recognizes 85 kDa NSF of mouse, and rat origins.
Target Species Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.