Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NSF Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $464.00
Specifications
Antigen | NSF |
---|---|
Concentration | 0.5 mg/ml |
Dilution | Western Blot 1 - 2 ug/mL, Immunohistochemistry 10 - 20 ug/mL, Immunoprecipitation 5 - 15 ug/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
NSF Polyclonal specifically detects NSF in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
NSF | |
Western Blot 1 - 2 ug/mL, Immunohistochemistry 10 - 20 ug/mL, Immunoprecipitation 5 - 15 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, 1% BSA, 50% Glycerol with 0.02% Sodium Azide | |
EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF | |
NSF | |
IgG | |
Affinity purified | |
The antibody recognizes 85 kDa NSF of mouse, and rat origins. |
0.5 mg/ml | |
Immunoprecipitation, Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
Q9QUL6 | |
4905 | |
Synthetic peptide corresponding to amino acids around 731 of rat NSF. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
83 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title