Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
nSMase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159936
Description
nSMase Polyclonal specifically detects nSMase in Human samples. It is validated for Western Blot.Specifications
| nSMase | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.1.4.12, ISC1, lyso-PAF-PLC, Lyso-platelet-activating factor-phospholipase C, Neutral sphingomyelinase, N-SMase, NSMASE, NSMASE1, sphingomyelin phosphodiesterase 2, sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O60906 | |
| SMPD2 | |
| Synthetic peptides corresponding to SMPD2(sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)) The peptide sequence was selected from the N terminal of SMPD2. Peptide sequence MKPNFSLRLRIFNLNCWGIPYLSKHRADRMRRLGDFLNQESFDLALLEEV The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Lipid and Metabolism | |
| 6610 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction