Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
nSMase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | nSMase |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
nSMase Polyclonal specifically detects nSMase in Human samples. It is validated for Western Blot.Specifications
| nSMase | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| 6610 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O60906 | |
| SMPD2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title