Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSP 5 alpha 3 alpha Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB100518C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100518C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100518C Supplier Novus Biologicals Supplier No. NB100518C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NSP 5 alpha 3 alpha Polyclonal specifically detects NSP 5 alpha 3 alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen NSP 5 alpha 3 alpha
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q5M775
Gene Alias cytospin-B, CYTSBcytospin B, FLJ36955, HCMOGT-1, NSP, NSP5, Nuclear structure protein 5, sperm antigen HCMOGT 1, Sperm antigen HCMOGT-1, sperm antigen with calponin homology and coiled coil domains 1, sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure protein NSP5b3b
Gene Symbols SPECC1
Host Species Rabbit
Immunogen An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 200-300). [UniProt# Q5M775]
Molecular Weight of Antigen 80 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 92521
Test Specificity Human. Immunogen has 93% homology with mouse and 87% homology with rat.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.