Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSP 5 alpha 3 alpha Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NB100517

 View more versions of this product

Catalog No. NB100517


Only null left
Add to Cart

Description

Description

NSP 5 alpha 3 alpha Polyclonal specifically detects NSP 5 alpha 3 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

NSP 5 alpha 3 alpha
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
cytospin-B, CYTSBcytospin B, FLJ36955, HCMOGT-1, NSP, NSP5, Nuclear structure protein 5, sperm antigen HCMOGT 1, Sperm antigen HCMOGT-1, sperm antigen with calponin homology and coiled coil domains 1, sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure protein NSP5b3b
Rabbit
80 kDa
0.1 mL
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience
92521
Human
IgG
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
1 mg/mL
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation
Q5M775
SPECC1
An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 1-100). [UniProt# Q5M775]
Affinity Purified
RUO
Primary
Human. Immunogen has 83% homology with mouse and rat.
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only