Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSP 5 alpha 3 alpha Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen NSP 5 alpha 3 alpha
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100517SS
SDP
View Documents
Novus Biologicals
NB100517SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB100517
SDP
View Documents
Novus Biologicals
NB100517
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

NSP 5 alpha 3 alpha Polyclonal specifically detects NSP 5 alpha 3 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

NSP 5 alpha 3 alpha
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation
Polyclonal
Rabbit
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
cytospin-B, CYTSBcytospin B, FLJ36955, HCMOGT-1, NSP, NSP5, Nuclear structure protein 5, sperm antigen HCMOGT 1, Sperm antigen HCMOGT-1, sperm antigen with calponin homology and coiled coil domains 1, sperm antigen with calponin homology and coiled-coil domains 1cytokinesis and spindle organization B, sperm antigen with calponin-like and coiled coil domains 1, structure protein NSP5a3a, structure protein NSP5a3b, structure protein NSP5b3a, structure protein NSP5b3b
SPECC1
IgG
Affinity Purified
Human. Immunogen has 83% homology with mouse and rat.
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
RUO
Human
Q5M775
92521
An internal synthetic peptide made to the human NSP 5 alpha 3 alpha protein sequence (between residues 1-100). [UniProt# Q5M775]
Primary
Store at 4C. Do not freeze.
80 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.