Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nsr1p Antibody (31C4), Novus Biologicals™
SDP

Catalog No. NB018367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018367 0.125 mL
NBP105389 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018367 Supplier Novus Biologicals Supplier No. NBP1053890.125ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nsr1p Monoclonal specifically detects Nsr1p in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nsr1p
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 31C4
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500
Formulation Tissue culture supernatant with 5mM Sodium Azide
Gene Alias NSR 1, Nsr 1p, NSR1, Nuclear localization sequence binding protein, p67, SHE 5, SHE5
Host Species Mouse
Immunogen Yeast nuclear preparation.
Molecular Weight of Antigen 44.5 kDa
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.