Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nsr1p Antibody (31C4), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.90

Specifications

Antigen Nsr1p
Clone 31C4
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018367
SDP
View Documents
Novus Biologicals
NBP1053890.125ML
0.125 mL
Each for $181.90
Only null left
Add to Cart
 
NBP105389
SDP
View Documents
Novus Biologicals
NBP105389
0.5 mL N/A N/A N/A
Description

Description

Nsr1p Monoclonal specifically detects Nsr1p in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nsr1p
Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500
Monoclonal
Supernatant
RUO
NSR 1, Nsr 1p, NSR1, Nuclear localization sequence binding protein, p67, SHE 5, SHE5
IgG1
Tissue culture supernatant
44.5 kDa
31C4
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Mouse
Yeast
Yeast nuclear preparation.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.