Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSRP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB343995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343995 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343995 Supplier Novus Biologicals Supplier No. NBP241139MFV450

Chicken Polyclonal Antibody

NSRP1 Polyclonal antibody specifically detects NSRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NSRP1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CCDC55, coiled-coil domain containing 55, coiled-coil domain-containing protein 55, DKFZp434K1421, FLJ37233, HSPC095, NSRP70, nuclear speckle splicing regulatory protein 1
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human CCDC55. The immunogen is located within amino acids 430 - 480 of CCDC55. Amino Acid Squence: VQSSERNQDRKESSPN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84081
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.