Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT-proBNP Antibody (24E11cc), Novus Biologicals™
SDP

Catalog No. NB12013123 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12013123 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12013123 Supplier Novus Biologicals Supplier No. NB12013123
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, Sandwich ELISA.

Specifications

Antigen NT-proBNP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2400000000000
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000
Gene Alias BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Gene Symbols NPPB
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 61-76, conjugated with carrier protein.
Purification Method Protein A purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4879
Test Specificity Human NT-proBNP and proBNP
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.