Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT-proBNP Antibody (OTI11G4), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP271213C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271213C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271213C Supplier Novus Biologicals Supplier No. NBP271213C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NT-proBNP Monoclonal specifically detects NT-proBNP in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NT-proBNP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI11G4
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Gene Symbols NPPB
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 27-102 of human BNP (NP_002512) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4879.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.