Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT2NL Antibody, Novus Biologicals™
SDP

Catalog No. NBP219595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219595 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219595 Supplier Novus Biologicals Supplier No. NBP219595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NT2NL Polyclonal specifically detects NT2NL in Human samples. It is validated for Western Blot.

Specifications

Antigen NT2NL
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q7Z3S9
Gene Alias FLJ35032, FLJ55807, N2NNotch homolog 2 (Drosophila) N-terminal like, notch 2 N-terminal like, Notch homolog 2 N-terminal like protein, notch homolog 2 N-terminal-like protein
Gene Symbols NOTCH2NL
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NOTCH2NL. The exact sequence is proprietary.
Molecular Weight of Antigen 26 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 388677
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.