Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5C1A Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB180593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180593 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180593 Supplier Novus Biologicals Supplier No. NBP306431V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NT5C1A Polyclonal antibody specifically detects NT5C1A in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NT5C1A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 5'-nucleotidase, cytosolic IA, AMP-specific 5'-NT, CN1, cN1A, CN-I, cN-IA, cytosolic 5' nucleotidase, type 1A, cytosolic 5'-nucleotidase 1A, Cytosolic 5'-nucleotidase IA, EC 3.1.3.5, MGC119199, MGC119201
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NT5C1A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84618
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.