Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5C1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156911
Description
NT5C1B Polyclonal specifically detects NT5C1B in Human, Bovine samples. It is validated for Western Blot.Specifications
NT5C1B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
5'-nucleotidase, cytosolic IB, AIRPcytosolic 5'-nucleotidase 1B, Autoimmune infertility-related protein, CN1B, cN-IB, Cytosolic 5'-nucleotidase IB, EC 3.1.3.5, MGC26640 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Mouse: 78%; Rat: 78%. | |
Human, Bovine, Rat, Rabbit, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B7ZVX7 | |
NT5C1B | |
Synthetic peptides corresponding to NT5C1B(5'-nucleotidase, cytosolic IB) The peptide sequence was selected from the N terminal of NT5C1B. Peptide sequence MSQTSLKQKKNEPGMRSSKESLEAEKRKESDKTGVRLSNQGSQESSLRKT. | |
100 μL | |
Lipid and Metabolism | |
93034 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction