Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NT5C1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NT5C1B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NT5C1B Polyclonal specifically detects NT5C1B in Human, Bovine samples. It is validated for Western Blot.Specifications
NT5C1B | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
5'-nucleotidase, cytosolic IB, AIRPcytosolic 5'-nucleotidase 1B, Autoimmune infertility-related protein, CN1B, cN-IB, Cytosolic 5'-nucleotidase IB, EC 3.1.3.5, MGC26640 | |
NT5C1B | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
B7ZVX7 | |
93034 | |
Synthetic peptides corresponding to NT5C1B(5'-nucleotidase, cytosolic IB) The peptide sequence was selected from the N terminal of NT5C1B. Peptide sequence MSQTSLKQKKNEPGMRSSKESLEAEKRKESDKTGVRLSNQGSQESSLRKT. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title