Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTAL Antibody (440005), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB370905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370905 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370905 Supplier Novus Biologicals Supplier No. FAB4066X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NTAL Monoclonal specifically detects NTAL in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen NTAL
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 440005
Conjugate Alexa Fluor 532
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5
Host Species Mouse
Immunogen E. coli-derived recombinant human NTAL, Met1-Ala243, Accession # Q9GZY6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7462
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.