Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTAL Antibody (NAP-07), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB500341Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500341Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500341Z Supplier Novus Biologicals Supplier No. NB500341AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NTAL Monoclonal specifically detects NTAL in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NTAL
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone NAP-07
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Gene Alias LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5
Gene Symbols LAT2
Host Species Mouse
Immunogen Recombinant cytoplasmic domain (amino acids 91-244) of Human NTAL.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7462
Test Specificity The antibody NAP-07 reacts with Non-T cell activation linker (NTAL), also known as LAB (linker of activated B cells), a 25 - 30 kDa transmembrane adaptor protein present in membrane microdomains (rafts) of B lymphocytes, NK cells and myeloid cells.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.