Learn More
Description
Specifications
Specifications
| Antigen | NTAL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.62mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5 |
| Gene Symbols | LAT2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
