Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTAN1 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB054195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054195 Supplier Novus Biologicals Supplier No. NBP298491H

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

NTAN1 Polyclonal specifically detects NTAN1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NTAN1
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias DKFZp666E058, EC 3.5.1.-, N-terminal asparagine amidase, N-terminal asparagine amidohydrolase, PNAA, PNAD, Protein NH2-terminal asparagine amidohydrolase, Protein NH2-terminal asparagine deamidase, Protein N-terminal Asn amidase, protein N-terminal asparagine amidohydrolase, Protein NTN-amidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NTAN1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 123803
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.