Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTB-A/SLAMF6/CD352 Antibody (110), Alexa Fluor™ 594
SDP

Catalog No. NB069124 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069124 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069124 Supplier Novus Biologicals Supplier No. NBP290029AF594

Rabbit Monoclonal Antibody

NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Alexa Fluor 594
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human NTB-A/SLAMF6/CD352 (Q96DU3-1; Met1-Met226).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 114836
Test Specificity No cross-reactivity in ELISA with: Human SLAMF7
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.