Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTB-A/SLAMF6/CD352 Antibody (13G3-19D), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP143344A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP143344A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP143344A70 Supplier Novus Biologicals Supplier No. NBP143344AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 13G3-19D
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Gene Symbols SLAMF6
Host Species Mouse
Immunogen The immunogen for this antibody was Ly108.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 114836.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.