Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTB-A/SLAMF6/CD352 Antibody (13G3-19D), Biotin, Novus Biologicals™
SDP

Catalog No. NBP14357801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14357801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP14357801 Supplier Novus Biologicals Supplier No. NBP1435780.1MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Flow Cytometry
Classification Monoclonal
Clone 13G3-19D
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Alias Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Gene Symbols SLAMF6
Host Species Mouse
Immunogen The immunogen for this antibody was Ly108.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114836
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.