Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTH1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB100108C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100108C 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100108C Supplier Novus Biologicals Supplier No. NB100108C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NTH1 Polyclonal specifically detects NTH1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen NTH1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P78549
Gene Alias EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1
Gene Symbols NTHL1
Host Species Rabbit
Immunogen A peptide derived from human NTH1, conjugated to KLH. [UniProt# P78549]
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4913
Test Specificity Human, mouse and rat. Immunogen sequence has 93% identity with bovine proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.