Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTNG1 Antibody (RM0292-4K62), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP211810UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211810UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211810UV Supplier Novus Biologicals Supplier No. NBP211810UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

NTNG1 Monoclonal specifically detects NTNG1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen NTNG1
Applications Western Blot
Classification Monoclonal
Clone RM0292-4K62
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias axon guidance molecule, KIAA0976YLSR571, laminet 1, laminet-1, Lmnt1, netrin G1, netrin G1f, netrin-G1
Gene Symbols NTNG1
Host Species Rat
Immunogen Mouse recombinant NTNG1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 22854
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.