Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NTPCR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238724
Description
NTPCR Polyclonal specifically detects NTPCR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NTPCR | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9BSD7 | |
NTPCR | |
This antibody was developed against a recombinant protein corresponding to amino acids: PVDGFYTEEVRQGGRRIGFDVVTLSGTRGPLSRVGLEPPPGKRECRVGQYVVDLTSFEQLALPV | |
0.1 mL | |
Protein Kinase | |
84284 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C1orf57, chromosome 1 open reading frame 57, EC 3.6.1.15, FLJ11383, HCR-NTPase, human cancer-related NTPase, MGC13186, NTPase, Nucleoside triphosphate phosphohydrolase, nucleoside-triphosphatase C1orf57, nucleoside-triphosphatase, cancer-related, RP4-678E16.2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction