Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTS1/NTSR1 Mouse anti-Human, Clone: B-N6, Novus Biologicals™
SDP

Catalog No. NB158400 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158400 500 μg
NB158399 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB158400 Supplier Novus Biologicals Supplier No. NBP314600500UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NTS1/NTSR1 Monoclonal antibody specifically detects NTS1/NTSR1 in Human samples. It is validated for Flow Cytometry, Functional Assay

Specifications

Antigen NTS1/NTSR1
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone B-N6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Functional
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias NTR
Host Species Mouse
Immunogen NTR transfected CHO cell line
Purification Method Precipitated in water
Quantity 500 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4923
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.