Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTS2/NTSR2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB003615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003615 Supplier Novus Biologicals Supplier No. NB10056472AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NTS2/NTSR2 Polyclonal specifically detects NTS2/NTSR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen NTS2/NTSR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias neurotensin receptor 2, neurotensin receptor type 2, neurotensin receptor, type 2, NT-R-2, NTR2Levocabastine-sensitive neurotensin receptor
Gene Symbols NTSR2
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Neurotensin receptor 2 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.