Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUB1/NYREN18 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB043521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043521 Supplier Novus Biologicals Supplier No. NBP297440B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NUB1/NYREN18 Polyclonal specifically detects NUB1/NYREN18 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NUB1/NYREN18
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias BS4, NEDD8 ultimate buster-1, negative regulator of ubiquitin-like proteins 1NUB1L, NY-REN-18 antigen, NYREN18NEDD8 ultimate buster 1, Renal carcinoma antigen NY-REN-18
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NUB1/NYREN18 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51667
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.