Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear Antigen Antibody (235-1R), PE, Novus Biologicals™
SDP

Catalog No. NB179735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179735 Supplier Novus Biologicals Supplier No. NBP314102PE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry

Specifications

Antigen Nuclear Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone 235-1R
Conjugate PE
Formulation PBS
Gene Alias Nuclei, Nucleus
Host Species Rabbit
Immunogen Nuclei of human myeloid leukemia biopsy cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.