Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear Antigen Antibody (rNM106) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB187402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187402 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187402 Supplier Novus Biologicals Supplier No. NBP314097
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Nuclear Antigen
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rNM106
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 to 2 μg /million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Formulation 10mM PBS without Preservative
Gene Alias Nuclei, Nucleus
Host Species Mouse
Immunogen Nuclei of HL60 cells
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.