Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Antigen Rabbit anti-Human, Primate, Clone: 235-1R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP313912100UG
This item is not returnable.
View return policy
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
Nuclear Antigen | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1000-2000 ug/500 ug protein lysate, Immunohistochemistry-Frozen 1-2 ug/ml | |
Nuclei, Nucleus | |
Nuclei of human myeloid leukemia biopsy cells | |
100 μg | |
Primary | |
Store at 4°C. | |
IgG κ |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
235-1R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction